Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRSS35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113205
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113205 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113205 Supplier Invitrogen™ Supplier No. PA5113205
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRSS35 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The PRSS35 (Protease, Serine 35) gene encodes a protein that belongs to the trypsin-like serine protease family, which is involved in various physiological processes, including digestion, immune response, and blood coagulation. PRSS35 is located on chromosome 6 in humans. The protein is believed to play a role in extracellular matrix remodeling and may be involved in processes such as tissue repair and inflammation. Expression of PRSS35 has been detected in multiple tissues, including the reproductive system, where it may influence fertility and reproductive tract function. However, the specific substrates and detailed biological functions of PRSS35 are still under investigation. Alterations in PRSS35 expression have been associated with certain pathological conditions, such as cancer and inflammatory diseases, suggesting its potential as a biomarker and therapeutic target.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS35
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PRSS35
Gene Accession No. Q8C0F9, Q8N3Z0
Gene Alias 6030424L22Rik; C6orf158; dJ223E3.1; Inactive serine protease 35; P3D9; protease, serine 35; protease, serine, 35; PRSS35; UNQ522/PRO1057
Gene Symbols PRSS35
Host Species Rabbit
Immunogen Recombinant Human Inactive serine protease 35 protein (201-413AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167681, 244954
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.