Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRSS37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554229 Supplier Invitrogen™ Supplier No. PA554229
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQDDLMLIKL AKPAMLNPKV QPLTLATTNV RPGTVCLLSG LDWSQENSGR HPDLRQNLEA PVMSDREC Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.

PRSS37 plays a role in male fertility. May have a role in sperm migration or binding to zona-intact eggs. Involved in the activation of the proacrosin/acrosin system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS37
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRSS37
Gene Accession No. A4D1T9
Gene Alias 1700016G05Rik; LOC540062 protein; Peptidase S1 domain-containing protein LOC136242; probable inactive serine protease 37; Probable inactive trypsin-X2; protease, serine 37; protease, serine, 37; Prss37; RGD1311823; serine protease 37; Tryx2
Gene Symbols PRSS37
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRSS37. Recombinant protein control fragment (Product #RP-92860).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136242
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.