Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRSS38 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551199
Description
The antibody detects endogenous levels of total PRSS38 protein.
The function of this protein remains unknown.
Specifications
PRSS38 | |
Polyclonal | |
Unconjugated | |
PRSS38 | |
marapsin 2; marapsin-2; MPN2; protease, serine 38; protease, serine, 38; PRSS38; Serine protease 38; serine protease MPN2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339501 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
A1L453 | |
PRSS38 | |
Full length fusion protein of human PRSS38. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction