Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRSS38 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563186
Description
Immunogen sequence: IYDMYVGLVN LRVAGNHTQW YEVNRVILHP TYEMYHPIGG DVALVQLKTR IVFSESVLPV CLATPE Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 64%.
The function of this protein remains unknown.
Specifications
PRSS38 | |
Polyclonal | |
Unconjugated | |
PRSS38 | |
marapsin 2; marapsin-2; MPN2; protease, serine 38; protease, serine, 38; PRSS38; Serine protease 38; serine protease MPN2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339501 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A1L453 | |
PRSS38 | |
Recombinant protein corresponding to Human PRSS38. Recombinant protein control fragment (Product #RP-103953). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction