Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRSS44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571245 Supplier Invitrogen™ Supplier No. PA571245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%; Sheep: 86%; Zebrafish: 93%.

PRSS44 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS44
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRSS44
Gene Accession No. E7EML9
Gene Alias 1700036D21Rik; 79H19A; AU043574; AU043597; protease, serine 44; protease, serine, 44; Prss44; RGD1560940; Serine protease 44; serine protease-like 1; Tessp4; TESSP-4; testis serine protease 4; testis specific serine protease 4; testis-specific serine protease 4
Gene Symbols PRSS44
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human PRSS44.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729756
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.