Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRSS54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572124
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572124 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572124 Supplier Invitrogen™ Supplier No. PA572124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRSS54 Polyclonal Antibody for Western Blot

This gene encodes a putative serine-type endopeptidase containing the peptidase S1 domain. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS54
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PRSS54
Gene Accession No. Q6PEW0, Q7M756
Gene Alias 4931432M23Rik; Cancer/testis antigen 67; CT67; Inactive serine protease 54; Klkbl4; Plasma kallikrein-like protein 4; protease, serine 54; protease, serine, 54; PRSS54; testis tissue sperm-binding protein Li 40a
Gene Symbols PRSS54
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 339-367 amino acids from the C-terminal region of human KLKBL4.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221191, 70993
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.