Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRSS55 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546604
Description
Peptide sequence: SWGKSCGEKN TPGIYTSLVN YNLWIEKVTQ LEGRPFNAEK RRTSVKQKPM Sequence homology: Human: 100%.
This gene encodes a member of a group of membrane-anchored chymotrypsin (S1)-like serine proteases. The enocoded protein is primarily expressed in the Leydig and Sertoli cells of the testis and may be involved in male fertility. Alternate splicing results in multiple transcript variants.
Specifications
PRSS55 | |
Polyclonal | |
Unconjugated | |
PRSS55 | |
CT153; probable serine protease UNQ9391/PRO34284; protease, serine 55; protease, serine, 55; PRSS55; Serine protease 55; Testis serine protease 1; tryptophan/serine protease; TSP1; T-SP1; UNQ9391; UNQ9391/PRO34284 | |
Rabbit | |
Affinity chromatography | |
RUO | |
203074 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6UWB4 | |
PRSS55 | |
Synthetic peptide directed towards the C-terminal of human PRSS55 (aa 271-320). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction