Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRSS58 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570559 Supplier Invitrogen™ Supplier No. PA570559
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Horse: 79%; Human: 100%; Rabbit: 79%.

This gene encodes a member of the trypsin family of serine proteases. This gene and several related trypsinogen genes are localized to the T cell receptor beta locus on chromosome 7. This gene was previously described as a trypsinogen-like pseudogene, but it is now thought to be a protein-coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS58
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRSS58
Gene Accession No. Q8IYP2
Gene Alias 1700020H15; LOC506919 protein; LOW QUALITY PROTEIN: serine protease 58; probable inactive serine protease 58; probable inactive trypsin-X3; protease, serine 58; protease, serine, 58; PRSS1; PRSS58; Putative inactive serine protease 58; putative trypsin-X3; serine protease 58; TRY1; trypsin X3; trypsin-X3; Tryx3; UNQ2540; UNQ2540/PRO6090
Gene Symbols PRSS58
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human PRSS58 (aa 122-171).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136541
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.