Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRSS8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987015
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7015 Supplier Bioss Supplier No. BS6064R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is highly expressed in prostate epithelia and is one of several proteolytic enzymes found in seminal fluid. The proprotein is cleaved to produce a light chain and a heavy chain which are associated by a disulfide bond. It is active on peptide linkages involving the carboxyl group of lysine or arginine.

Specifications

Antigen PRSS8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PRSS8
Gene Accession No. Q16651
Gene Alias 2410039E18Rik; AI313909; BOS_22900; C79772; CAP1; channel activating protease 1; Channel-activating protease 1; fr; mCAP1; Prostasin; Prostasin heavy chain; Prostasin light chain; protease, serine 8; protease, serine 8 (prostasin); protease, serine, 8; protease, serine, 8 (prostasin); Prss8; Serine protease 8
Gene Symbols PRSS8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PRSS8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5652
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.