Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PRT 4165
GREENER_CHOICESDP

Catalog No. 504750R Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
504750R 50 mg
504710R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 504750R Supplier R&D Systems™ Supplier No. 5047/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of Bmi1/Ring1A; blocks histone H2A ubiquitination

PRT 4165 is an inhibitor of Bmi1/Ring1A, subunits of the polycomb repressive complex 1 (PRC1); inhibits self-ubiquitination (IC50 = 3.9 μM) but does not increase cellular levels of either subunit. Prevents Bmi1/Ring1A-mediated ubiquitination and drug-induced degradation of topoisomerase 2α (Top2α). Also shown to inhibit the in vitro E3 ubiquitin ligase activity of RNF2 and a Bmi1/RNF2 complex, inhibiting H2A/H2AX ubiquitination. Blocks polycomb repressor complex (PRC) 1-mediated histone H2A ubiquitination in vitro.

Specifications

Quantity 50 mg
Product Line Tocris
Product Type PRT 4165
Purity >99%
Molecular Weight (g/mol) 235.24
Inhibitors PRT 4165

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.