Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRTFDC1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300245PE
Description
PRTFDC1 Polyclonal antibody specifically detects PRTFDC1 in Human samples. It is validated for Western Blot, ELISASpecifications
PRTFDC1 | |
Polyclonal | |
Western Blot, ELISA | |
EC 2.4.2.8, FLJ11888, HHGPEC 2.4.2, phosphoribosyl transferase domain containing 1, phosphoribosyltransferase domain-containing protein 1 | |
Produced in rabbits immunized with purified, recombinant Human PRTFDC1 (Uniprot#: Q9NRG1-1; Met1-Val225) | |
0.1 mL | |
Lipid and Metabolism | |
56952 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction