Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRTFDC1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520593

Catalog No. PIPA520593


Only null left
Add to Cart

Description

Description

A suggested positive control is human brain tissue lysate. PA5-20593 can be used with blocking peptide PEP-0713.

Phosphoribosyl transferase domain containing 1 (PRTFDC1) is highly homologous to the hypoxanthine phosphoribosyltransferase (HPRT1) and may have arisen from a gene duplication event of a common ancestor gene. Recently, it was shown that CpG islands in the PRTFDC1 promoter could be hypermethylated in ovarian cancers and oral squamous-cell carcinomas (OSCC), leading to gene silencing. Restoration of PRTFDC1 expression in OSCC inhibited cell growth in colony-formation assays, while knockdown of PRTFDC1 expression in OSCC that expressed the gene promoted cell growth. These results suggest that PRTFDC1 can act as a tumor-suppressor gene. At least three isoforms of PRTFDC1 are known to exist.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRTFDC1
Polyclonal
Unconjugated
PRTFDC1
HHGP; phosphoribosyl transferase domain containing 1; phosphoribosyltransferase domain-containing protein 1; PRTFDC1
Rabbit
Antigen affinity chromatography
RUO
291355, 56952
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9NRG1
PRTFDC1
A 14 amino acid peptide from near the center of human PRTFDC1.
100 μg
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.