Learn More
PRTFDC1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305706AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Phosphoribosyl transferase domain containing 1 (PRTFDC1) is highly homologous to the hypoxanthine phosphoribosyltransferase (HPRT1) and may have arisen from a gene duplication event of a common ancestor gene. Recently, it was shown that CpG islands in the PRTFDC1 promoter could be hypermethylated in ovarian cancers and oral squamous-cell carcinomas (OSCC), leading to gene silencing. Restoration of PRTFDC1 expression in OSCC inhibited cell growth in colony-formation assays, while knockdown of PRTFDC1 expression in OSCC that expressed the gene promoted cell growth. These results suggest that PRTFDC1 can act as a tumor-suppressor gene. At least three isoforms of PRTFDC1 are known to exist.Specifications
PRTFDC1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NRG1 | |
PRTFDC1 | |
Between 175 and 225 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
PRTFDC1 | |
HHGP; phosphoribosyltransferase domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56952 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.