Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRUNE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555842
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555842 Supplier Invitrogen™ Supplier No. PA555842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LERSHSPPLK LTPASSTHPN LHAYLQGNTQ VSRKKLLPLL QEALSAYFDS MKIPSGQPET ADVSREQ Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 71%.

Phosphodiesterase (PDE) that has higher activity toward cAMP than cGMP, as substrate. It plays a role in cell proliferation, is able to induce cell motility and acts as a negative regulator of NME1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRUNE
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRUNE1
Gene Accession No. Q86TP1
Gene Alias 9230112O05Rik; C130058A12; DRES17; DRES-17; Drosophila-related expressed sequence 17; Exopolyphosphatase PRUNE1; hPrune; H-PRUNE; HTcD37; protein prune homolog; Protein prune homolog 1; Prune; prune exopolyphosphatase; prune homolog; prune homolog (Drosophila); PRUNE1; PRUNEM1; Prune-M1
Gene Symbols PRUNE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRUNE. Recombinant protein control fragment (Product #RP-94279).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58497
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.