Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRUNE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598564
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598564 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598564 Supplier Invitrogen™ Supplier No. PA598564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRUNE Polyclonal Antibody for ELISA, ICC/IF, IHC (P), IP, Western Blot

Phosphodiesterase (PDE) that has higher activity toward cAMP than cGMP, as substrate. It plays a role in cell proliferation, is able to induce cell motility and acts as a negative regulator of NME1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRUNE
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PRUNE1
Gene Accession No. Q86TP1
Gene Alias 9230112O05Rik; C130058A12; DRES17; DRES-17; Drosophila-related expressed sequence 17; Exopolyphosphatase PRUNE1; hPrune; H-PRUNE; HTcD37; protein prune homolog; Protein prune homolog 1; Prune; prune exopolyphosphatase; prune homolog; prune homolog (Drosophila); PRUNE1; PRUNEM1; Prune-M1
Gene Symbols PRUNE1
Host Species Rabbit
Immunogen Recombinant Human Exopolyphosphatase PRUNE1 protein (1-168AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58497
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.