Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRUNE2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PRUNE2 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PRUNE2 Polyclonal specifically detects PRUNE2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PRUNE2 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
158471 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LNDEGKLSITLVGSSVLASEDKTLESAVVKVINPVEQSDANVEFRESSSSLVLKEILQEAPELITEQLAHRLRGSILFKWMT | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
A214N16.3, bA214N16.3, BCH motif-containing molecule at the carboxyl terminal region 1, BMCC1FLJ50060, BNIP2 motif containing molecule at the carboxyl terminal region 1, BNIP2 motif-containing molecule at the C-terminal region 1, BNIPXLFLJ59118, C9orf65, chromosome 9 open reading frame 65, DKFZp762K117, KIAA0367FLJ54876, protein prune homolog 2, prune homolog 2 (Drosophila), RP11-58J3.2 | |
PRUNE2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title