Learn More
Invitrogen™ PRUNE2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total PRUNE2.
Specifications
Specifications
| Antigen | PRUNE2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PRUNE2 |
| Gene Accession No. | Q52KR3, Q5BJR4, Q8WUY3 |
| Gene Alias | 6330414G02Rik; A214N16.3; A230083H22Rik; A330102H22Rik; bA214N16.3; BCH motif-containing molecule at the carboxyl terminal region 1; Bmcc1; BNIP2 motif containing molecule at the carboxyl terminal region 1; BNIP2 motif-containing molecule at the C-terminal region 1; BNIPXL; C9orf65; DKFZp762K117; KIAA0367; mKIAA0367; neuronal protein; olfaxin; protein prune homolog 2; prune homolog 2; prune homolog 2 (Drosophila); PRUNE2; RGD1311350; RP11-214N16.3; RP11-58J3.2; truncated PRUNE2 |
| Gene Symbols | PRUNE2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.