Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRX Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110225
Description
Immunogen sequence: LGARARVGGE GAEEQPPGAE RTFCLSLPDV ELSPSGGNHA EYQVAEGEGE AGHKLKVRLP RFGLVRAKEG AEEGEKAKSP KLRLPRVGFS QSEMVTGEGS P.
Periaxin is a protein that in humans is encoded by the PRX gene. This gene encodes a protein involved in peripheral nerve myelin upkeep. The encoded protein contains 2 PDZ domains which were named after PSD95 (post synaptic density protein), DlgA (Drosophila disc large tumor suppressor), and ZO1 (a mammalian tight junction protein). Two alternatively spliced transcript variants have been described for this gene which encode different protein isoforms and which are targeted differently in the Schwann cell. Mutations in this gene cause Charcot-Marie-Tooth neuoropathy, type 4F and Dejerine-Sottas neuropathy.
Specifications
| PRX | |
| Polyclonal | |
| Unconjugated | |
| PRX | |
| CMT4F; KIAA1620; L-Periaxin; periaxin; PRX | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19153, 57716, 78960 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.77 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O55103, Q63425, Q9BXM0 | |
| PRX | |
| A synthetic peptide corresponding to a sequence within amino acids 1250-1350 of human PRX (NP_870998.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction