Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551646 Supplier Invitrogen™ Supplier No. PA551646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKVPDMKLPE IKLPKVPEMA VPDVHLPEVQ LPKVSEIRLP EMQVPKVPDV HLPKAPEVKL PRAPEVQLKA TKAEQAEGME FGFKMPKMTM PKLGRAESPS RGKPGEAGAE VSGKLVTLPC LQPEVDGEAH VGVPSLTLPS VELDL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.

Periaxin is a protein that in humans is encoded by the PRX gene. This gene encodes a protein involved in peripheral nerve myelin upkeep. The encoded protein contains 2 PDZ domains which were named after PSD95 (post synaptic density protein), DlgA (Drosophila disc large tumor suppressor), and ZO1 (a mammalian tight junction protein). Two alternatively spliced transcript variants have been described for this gene which encode different protein isoforms and which are targeted differently in the Schwann cell. Mutations in this gene cause Charcot-Marie-Tooth neuoropathy, type 4F and Dejerine-Sottas neuropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRX
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRX
Gene Accession No. Q9BXM0
Gene Alias CMT4F; KIAA1620; L-Periaxin; periaxin; PRX
Gene Symbols PRX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRX. Recombinant protein control fragment (Product #RP-100995).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57716
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.