Learn More
Invitrogen™ PRY Polyclonal Antibody

Description
PRY Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PRY |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | PRY |
| Gene Accession No. | O14603 |
| Gene Alias | PRY; PRY1; PRY2; PRYP3; PRYP4; PTPN13-like protein, Y-linked; PTPN13-like, Y-linked; PTPN13-like, Y-linked 2; PTPN13-like, Y-linked, centromeric; PTPN13LY; PTPN13LY2; testis-specific PTP-BL-related protein on Y; testis-specific PTP-BL-related Y protein |
| Gene Symbols | PRY |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.