Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRY Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583814
Description
Immunogen sequence: ATGLGFLLSW RQDNLNGTDC QGCNILYFSE TTGSMCSELS LNRGLEARRK KDLKDSFLW.
This gene is located in the nonrecombining portion of the Y chromosome, and expressed specifically in testis. It encodes a protein which has a low degree of similarity to protein tyrosine phosphatase, non-receptor type 13. Two nearly identical copies of this gene exist within a palindromic region. This record represents the more telomeric copy.
Specifications
PRY | |
Polyclonal | |
Unconjugated | |
PRY | |
PRY; PRY1; PRY2; PRYP3; PRYP4; PTPN13-like protein, Y-linked; PTPN13-like, Y-linked; PTPN13-like, Y-linked 2; PTPN13-like, Y-linked, centromeric; PTPN13LY; PTPN13LY2; testis-specific PTP-BL-related protein on Y; testis-specific PTP-BL-related Y protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9081 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14603 | |
PRY | |
Recombinant protein corresponding to Human PRY. Recombinant protein control fragment (Product #RP-100073). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction