Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140317 Supplier Invitrogen™ Supplier No. PA5140317
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: YGKVGCISLP LREMTAWINP PQISEIFQGY HQRVHGADAL SLQTNSLRSR LSSQCLGQSF LLRTLERGRG FRAL Highest antigen sequence indentity to the following orthologs: Mouse - 28%, Rat - 25%.

PRY2 (PTPN13 Like Y-Linked 2) is a Protein Coding gene. Diseases associated with PRY2 include Spermatogenic Failure, Y-Linked, 2 and Y Chromosome Infertility. An important paralog of this gene is PRYP3. This gene is located in the nonrecombining portion of the Y chromosome, and expressed specifically in testis. It encodes a protein which has a low degree of similarity to protein tyrosine phosphatase, non-receptor type 13. Two nearly identical copies of this gene exist within a palindromic region. This record represents the more centromeric copy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRY
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRY2
Gene Accession No. O14603
Gene Alias PTPN13-Like Protein, Y-Linked; PTPN13LY; PTPN13LY2
Gene Symbols PRY2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRY2. Recombinant protein control fragment (Product #RP-108976).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442862
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.