Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PRY2 Recombinant Protein Antigen
SDP

Catalog No. NB171398 Shop All R&D Systems Products
Change view
Click to view available options
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NB171398 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171398 Supplier Novus Biologicals™ Supplier No. NBP317614PEP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 442862
Molecular Weight (g/mol) M.W. Theoretical: 26 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias PRY1, PTPN13-Like Protein, Y-Linked, PTPN13-Like, Y-Linked 2, PTPN13-Like, Y-Linked, Centromeric, PTPN13LY, PTPN13LY2, Testis-Specific PTP-BL-Related Protein On Y, Testis-Specific PTP-BL-Related Y Protein
Gene Symbol PRY2
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein PRY2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.