Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSA/Kallikrein 3 Polyclonal Antibody, Abbomax

Catalog No. 502203937
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3937 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3937 Supplier Abbomax Inc Supplier No. 600860
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: LNCaP cell lysate, prostate ,pros Cellular location: Cytoplasmic.

PSA/Kallikrein 3 Polyclonal antibody specifically detects PSA/Kallikrein 3 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PSA/Kallikrein 3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene KLK3
Gene Accession No. P07288
Gene Alias antigen, prostate specific; APS; Fletcher factor; gamma-seminoprotein; hK3; Kal3; Kal-3; kallikrein 3, plasma; kallikrein B, plasma 1; kallikrein related peptidase 3; kallikrein-3; kallikrein-related peptidase 3; Kalp1; KALP15; Kininogenin; KLK2A1; Klk3; KLK-3; Klkb1; P-30 antigen; Pk; Plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein; prostate specific antigen; prostate-specific antigen; PSA; Semenogelase; seminin
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-Terminus of human PSA protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 354
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.