Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ psaA Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144568 Supplier Invitrogen™ Supplier No. PA5144568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

psaA Polyclonal Antibody for Western Blot, ELISA

Part of the ATP-binding cassette (ABC) transport system PsaABC involved in manganese import (By similarity). Binds manganese with high affinity and specificity and delivers it to the membrane permease for translocation into the cytoplasm (By similarity). Also acts as an adhesin which is involved on adherence to extracellular matrix (By similarity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen psaA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene psaA
Gene Accession No. P42363
Gene Alias papA
Gene Symbols psaA
Host Species Rabbit
Immunogen Recombinant Streptococcus pneumoniae psaA protein (21-310aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66806726
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.