Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSAP Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | PSAP |
---|---|
Concentration | 0.94 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119361
|
Invitrogen™
PA5119361 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121667
|
Invitrogen™
PA5121667 |
20 μL |
Each for $209.50
|
|
|||||
Description
PSAP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
PSAP is a highly conserved glycoprotein which is a precursor for 4 cleavage products: saposins A, B, C, and D. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities.Specifications
PSAP | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Psap | |
A1 activator; AI037048; Cerebroside sulfate activator; Co-beta-glucosidase; Component C; CSAct; Dispersin; GLBA; Glucosylceramidase activator; Proactivator polypeptide; prosaposin; prosaposin (sulfated glycoprotein, sphingolipid hydrolase activator); Protein A; Protein C; Psap; SAP1; SAP-1; SAP-2; Saposin-A; Saposin-B; Saposin-B-Val; Saposin-C; Saposin-D; Sgp1; SGP-1; SGP1A; snoRNA MBII-198; Sphingolipid activator protein 1; Sphingolipid activator protein 2; sphingolipid activator protein-1; sulfated glycoprotein 1; Sulfatide/GM1 activator | |
Psap | |
Primary | |
Antigen affinity chromatography |
0.94 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P07602 | |
5660 | |
Recombinant protein Prosaposin. The antigen corresponds to amino acid range 17-524 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title