Learn More
Invitrogen™ PSAP Polyclonal Antibody

Description
Immunogen sequence: TNSTFVQALV EHVKEECDRL GPGMADICKN YISQYSEIAI QMMMHMQPKE ICALVGFCDE VKEMPMQTLV PAKVASKNVI PALELVEPIK KHEVPAKSDV YCEVCEFLVK EVTKLIDNNK TEKEILDAFD KMCSKLPKSL SEE Highest antigen sequence indentity to the following orthologs: Rat - 56%, Mouse - 51%.
Specifications
Specifications
| Antigen | PSAP |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Psap |
| Gene Accession No. | P07602 |
| Gene Alias | A1 activator; AI037048; Cerebroside sulfate activator; Co-beta-glucosidase; Component C; CSAct; Dispersin; GLBA; Glucosylceramidase activator; Proactivator polypeptide; prosaposin; prosaposin (sulfated glycoprotein, sphingolipid hydrolase activator); Protein A; Protein C; Psap; SAP1; SAP-1; SAP-2; Saposin-A; Saposin-B; Saposin-B-Val; Saposin-C; Saposin-D; Sgp1; SGP-1; SGP1A; snoRNA MBII-198; Sphingolipid activator protein 1; Sphingolipid activator protein 2; sphingolipid activator protein-1; sulfated glycoprotein 1; Sulfatide/GM1 activator |
| Gene Symbols | Psap |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.