Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PSAT1, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABC950MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABC950MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC950MI Supplier MilliporeSigma™ Supplier No. ABC950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phosphoserine aminotransferase (EC 2.6.1.52; UniProt Q9Y617; also known as Phosphohydroxythreonine aminotransferase, PSAT) is encoded by the PSAT1 (also known as NLS2, PSA, PSATD) gene (Gene ID 29968) in human. PSAT1 is a class-V pyridoxal-phosphate-dependent aminotransferase family member that catalyzes the reversible conversion of 3-phosphohydroxypyruvate and to 1-phosphoserine, as well as the conversion of 3-hydroxy-2-oxo-4-phosphonooxybutanoate to 4-phosphonooxy-L-threonine, using, in both cases, L-glutamate as an amine donor, which is converted to 2-oxoglutarate. PSAT1 is involved in serine biosynthesis from glycolytic intermediates and PSAT1 upregulation is reported in non-small cell lung cancer (NSCLC) and colon cancer cells, while PSAT1 knockdoown is shown to suppress the proliferation of MDA-MB-468 and BT-20 breast cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSAT1
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene PSAT1, NLS2, PSA, PSATD
Gene Accession No. Q9Y617
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence within the region between the two pyridoxal phosphate-binding domains of human PSAT1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_478059
Test Specificity The immunogen sequence is 100% conserved in murine PSAT1 (UniProt Q99K85) and present in both spliced isoforms of human PSAT1 (Uniprot Q9Y617).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.