Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSCA Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298728F
Description
PSCA Polyclonal antibody specifically detects PSCA in Human samples. It is validated for Western BlotSpecifications
PSCA | |
Polyclonal | |
Western Blot | |
PRO232, prostate stem cell antigen | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human PSCA. | |
0.1 mL | |
Cancer, Cellular Markers, Prostate Cancer, Stem Cell Markers | |
8000 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction