Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSCA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$81.70 - $330.00
Specifications
Antigen | PSCA |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PSCA Polyclonal antibody specifically detects PSCA in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
PSCA | |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cellular Markers, Prostate Cancer, Stem Cell Markers | |
0.2um filtered solution in PBS | |
8000 | |
IgG | |
Antigen and protein A Affinity-purified |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
PRO232, prostate stem cell antigen | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PSCA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title