Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSCA Monoclonal Antibody (C1), Invitrogen™

Catalog No. PIMA543623
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543623 20 μL
PIMA543622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543623 Supplier Invitrogen™ Supplier No. MA543623
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSCA (Prostate Stem Cell antigen) is a cell surface antigen, which is overexpressed in approximately40% of primary prostate cancers and in as many as 100% of metastatic ones. PSCA is also overexpressed in a majority of transitional cell and pancreatic carcinomas. Antibody directed against PSCA inhibits tumorigenesis, slows tumor growth, prolongs survival and prevents metastasis in a preclinical zenograft model indicating that PSCA may have utility as a prognostic marker and/or therapeutic target in prostate cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSCA
Applications Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PSCA
Gene Accession No. O43653
Gene Alias 2210408B04Rik; caldesmon; CDM; HCAD; H-CAD; LCAD; L-CAD; NAG22; PRO232; prostate stem cell antigen; PSCA; UNQ206; UNQ206/PRO232
Gene Symbols PSCA
Host Species Mouse
Immunogen Leu13-Ser86 with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8000
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.