Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSCD1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6707R

Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.Specifications
PSCD1 | |
Polyclonal | |
Unconjugated | |
CYTH1 | |
B2-1; CLM1; CTH-1; Cyth1; CYTIP; cytoadhesin 1; cytohesin 1; Cytohesin-1; D17S811E; FLJ34050; FLJ41900; homolog of secretory protein SEC7; mKIAA4240; mSec7-1; PH, SEC7 and coiled-coil domain-containing protein 1; pleckstrin homology, Sec7 and coiled/coil domains 1(cytohesin 1); pleckstrin homology, Sec7 and coiled-coil domains 1; Pscd1; rSec7-1; Sec7; SEC7 homolog A; SEC7 homolog B2-1; Sec7a | |
Rabbit | |
Protein A | |
RUO | |
9267 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q15438 | |
CYTH1 | |
KLH conjugated synthetic peptide derived from human PSCD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction