Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSD-95 Antibody (6G6-1C9), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300556MFV450
Description
PSD-95 Monoclonal antibody specifically detects PSD-95 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Flow CytometrySpecifications
PSD-95 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Invertebrate, Primate | |
Purified |
Flow Cytometry | |
6G6-1C9 | |
Flow Cytometry | |
discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15 | |
Purified recombinant rat PSD-95. | |
0.1 mL | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
1742 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction