Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSD-95 Antibody (6G6-1C9), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300556PE
Description
PSD-95 Monoclonal specifically detects PSD-95 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Flow Cytometry.Specifications
PSD-95 | |
Monoclonal | |
PE | |
discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Detects Post Synaptic Density 95 kDa (PSD-95) from rat tissues. | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
6G6-1C9 | |
Flow Cytometry | |
DLG4 | |
Purified recombinant rat PSD-95. | |
0.1 mL | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
1742 | |
Human, Mouse, Rat, Invertebrate, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction