Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSD-95 Antibody (OTI12A5), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273679AF350
Description
PSD-95 Monoclonal specifically detects PSD-95 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PSD-95 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
DLG4 | |
Full length recombinant protein of human PSD-95 (NP_001356) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI12A5 | |
Western Blot | |
discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15 | |
Mouse | |
Affinity Purified | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
1742.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction