Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSD-95 Rabbit anti-Mouse, Rat, Clone: 0M7N4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PSD-95 |
---|---|
Clone | 0M7N4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PSD-95 Monoclonal antibody specifically detects PSD-95 in Mouse, Rat samples. It is validated for Western BlotSpecifications
PSD-95 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1742 | |
IgG | |
Affinity purified |
0M7N4 | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Rat | |
discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PSD-95 (P78352). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title