Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563690 Supplier Invitrogen™ Supplier No. PA563690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRLEASTSDP LPARGGSALP GSRNLVHGPP APPQVGADGL YSSLPNGLGG PPERLATLFG GPADTGFLNQ GDTWSSPREV SSHAQRIARA KWE Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.

PSD, which called EFA6A, is 63% identical to EFA6B (PSD4) and contains a Sec7 domain adjacent to a PH domain, followed by a C-terminal coiled-coil motif. This gene is mapped to chromosome 10q24. And it encodes a Plekstrin homology and SEC7 domains-containing protein that functions as a guanine nucleotide exchange factor. The encoded protein regulates signal transduction by activating ADP-ribosylation factor 6. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSD
Gene Accession No. A5PKW4
Gene Alias 1110007H17Rik; A930015K15; EFA6; EFA6A; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6; Exchange factor for ARF6; Exchange factor for ARF6 A; KIAA2011; mKIAA2011; PH and SEC7 domain-containing protein 1; pleckstrin and Sec7 domain containing; pleckstrin and Sec7 domain containing homolog; pleckstrin homology and SEC7 domain-containing protein 1; pleckstrin-Sec7 domains-containing; Psd; Psd1; Psdl; TYL
Gene Symbols PSD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSD. Recombinant protein control fragment (Product #RP-100892).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.