Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598091
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598091 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598091 Supplier Invitrogen™ Supplier No. PA598091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSD Polyclonal Antibody for Western Blot, IHC (P), ELISA

PSD, which called EFA6A, is 63% identical to EFA6B (PSD4) and contains a Sec7 domain adjacent to a PH domain, followed by a C-terminal coiled-coil motif. This gene is mapped to chromosome 10q24. And it encodes a Plekstrin homology and SEC7 domains-containing protein that functions as a guanine nucleotide exchange factor. The encoded protein regulates signal transduction by activating ADP-ribosylation factor 6. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PSD
Gene Accession No. A5PKW4
Gene Alias 1110007H17Rik; A930015K15; EFA6; EFA6A; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6; Exchange factor for ARF6; Exchange factor for ARF6 A; KIAA2011; mKIAA2011; PH and SEC7 domain-containing protein 1; pleckstrin and Sec7 domain containing; pleckstrin and Sec7 domain containing homolog; pleckstrin homology and SEC7 domain-containing protein 1; pleckstrin-Sec7 domains-containing; Psd; Psd1; Psdl; TYL
Gene Symbols PSD
Host Species Rabbit
Immunogen Recombinant Human PH and SEC7 domain-containing protein 1 protein (247-489AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5662
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.