Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589821 Supplier Invitrogen™ Supplier No. PA589821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEEDKLLSAV PEEGDATRDP GPEPEEEPGV RNGMASEGLN SSLCSPGHER RGTPADTEEP TKDPDVAFHG LSLGLSLTNG LALGPDLNIL EDSAESRPWR AGVLAEGDNA SRSLYPDAED PQLGLDGPGE PDVRDGFSAT FEKILESELL RGTQYSSLDS LDGLSLTDES DSCVSFEAPL TPLIQQRARD; Positive Samples: U-87MG, U-251MG, Mouse brain, Rat brain; Cellular Location: Membrane, Single-pass membrane protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Psd2
Gene Accession No. Q6P1I6, Q9BQI7
Gene Alias 6330404E20Rik; AW125584; Efa6c; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6 C; Exchange factor for ARF6 C; PH and SEC7 domain-containing protein 2; pleckstrin and Sec7 domain containing 2; pleckstrin homology and SEC7 domain-containing protein 2; PSD2
Gene Symbols Psd2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human PSD2 (NP_1156651).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307500, 74002, 84249
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.