Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSD3 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB271015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271015 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271015 Supplier Novus Biologicals Supplier No. NBP318437AF750
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSD3 Polyclonal antibody specifically detects PSD3 in Human samples. It is validated for Western Blot

Specifications

Antigen PSD3
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ADP-ribosylation factor guanine nucleotide factor 6, DKFZp761K1423, EFA6RHepatocellular carcinoma-associated antigen 67, Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6, HCA67Pleckstrin homology and SEC7 domain-containing protein 3, KIAA0942, PH and SEC7 domain-containing protein 3, pleckstrin and Sec7 domain containing 3
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 998 and 1048..
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23362
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.