Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564885 Supplier Invitrogen™ Supplier No. PA564885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FSAPPFPAAIG SQKKFSRPLL PATTTKLSQE EQLKSHESKL KQITTELAEH RSYPPDKKVK AKDVDEYKLK DHYLEFEKTR YEMYVSILKE GGKELLSNDE SEAAGLKKSH SSPSLNPDTS PITAKVKRNV SERKDHRPET PSIKQ Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

Guanine nucleotide exchange factor for ARF6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSD3
Gene Accession No. Q9NYI0
Gene Alias 4931420C21Rik; ADP-ribosylation factor guanine nucleotide factor 6; AI661273; BC003498; D430018P08; EFA6D; EFA6R; Epididymis tissue protein Li 20mP; exchange factor for ADP-ribosylation factor guanine nucleotide factor 6; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6 D; Exchange factor for ARF6 D; guanine nucleotide exchange factor for ADP ribosylation factor 6; HCA67; Hepatocellular carcinoma-associated antigen 67; KIAA0942; PH and SEC7 domain-containing protein 3; pleckstrin and Sec7 domain containing 3; pleckstrin homology and SEC7 domain-containing protein 3; Psd3; RGD1559968
Gene Symbols PSD3
Host Species Rabbit
Immunogen Recombinant Human PSD3. Recombinant protein control fragment (Product #RP-105688).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23362
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.