Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSD3 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570895
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0895 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0895 Supplier Bethyl Laboratories, Inc Supplier No. A305625AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Guanine nucleotide exchange factor for ARF6.

Specifications

Antigen PSD3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PSD3
Gene Accession No. Q9NYI0
Gene Alias EFA6R, HCA67, ADP-ribosylation factor guanine nucleotide factor 6, PH and SEC7 domain-containing protein 3, exchange factor for ADP-ribosylation factor guanine nucleotide factor 6, hepatocellular carcinoma-associated antigen 67, pleckstrin homology and SEC7 domain-containi
Gene Symbols PSD3
Host Species Rabbit
Immunogen Between 998 and 1048
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23362
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.