Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110227
Description
Immunogen sequence: QTQSPEEMQG WINKINCVAA VFSAPPFPAA IGSQKKFSRP LLPATTTKLS QEEQLKSHES KLKQITTELA EHRSYPPDKK VKAKDVDEYK LKDHYLEFEK TRYEMYVSIL KEGGKELLSN DESEAAGLKK SHSSPSLNPD TSPITAKVKR NVSERKDHRP ETPSIKQKVT.
Guanine nucleotide exchange factor for ARF6.
Specifications
PSD3 | |
Polyclonal | |
Unconjugated | |
PSD3 | |
4931420C21Rik; ADP-ribosylation factor guanine nucleotide factor 6; AI661273; BC003498; D430018P08; EFA6D; EFA6R; Epididymis tissue protein Li 20mP; exchange factor for ADP-ribosylation factor guanine nucleotide factor 6; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6 D; Exchange factor for ARF6 D; guanine nucleotide exchange factor for ADP ribosylation factor 6; HCA67; Hepatocellular carcinoma-associated antigen 67; KIAA0942; PH and SEC7 domain-containing protein 3; pleckstrin and Sec7 domain containing 3; pleckstrin homology and SEC7 domain-containing protein 3; Psd3; RGD1559968 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
234353 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.86 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q2PFD7 | |
PSD3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 878-1047 of human PSD3 (NP_0561253). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction