Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557029 Supplier Invitrogen™ Supplier No. PA557029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EESMFFSNPL FLASPCSENS ASGECFSWGA SDSHAGVRTG PESPATLEPP LPEDTVLWEL ESEPDLGDGA AISGHCTPPF PVPIYKPHSI Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 55%.

Guanine nucleotide exchange factor for ARF6 and ARL14/ARF7. Through ARL14 activation, controls the movement of MHC class II-containing vesicles along the actin cytoskeleton in dendritic cells. Involved in membrane recycling. Interacts with several phosphatidylinositol phosphate species, including phosphatidylinositol 3,4-bisphosphate, phosphatidylinositol 3,5-bisphosphate and phosphatidylinositol 4,5-bisphosphate. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSD4
Gene Accession No. Q8NDX1
Gene Alias ADP-ribosylation factor guanine nucleotide-exchange factor 6; EFA6B; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6 B; exchange factor for ADP-ribosylation factor guanine nucleotide factor 6B; exchange factor for ARF6 B; PH and SEC7 domain-containing protein 4; pleckstrin and Sec7 domain containing 4; pleckstrin homology and SEC7 domain-containing protein 4; PSD4; SEC7 homolog; telomeric of interleukin-1 cluster protein; TIC
Gene Symbols PSD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSD4. Recombinant protein control fragment (Product #RP-96071).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.