Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599856
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599856 Supplier Invitrogen™ Supplier No. PA599856
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PSD4.

Guanine nucleotide exchange factor for ARF6 and ARL14/ARF7. Through ARL14 activation, controls the movement of MHC class II-containing vesicles along the actin cytoskeleton in dendritic cells. Involved in membrane recycling. Interacts with several phosphatidylinositol phosphate species, including phosphatidylinositol 3,4-bisphosphate, phosphatidylinositol 3,5-bisphosphate and phosphatidylinositol 4,5-bisphosphate. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PSD4
Gene Accession No. Q8NDX1
Gene Alias ADP-ribosylation factor guanine nucleotide-exchange factor 6; EFA6B; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6 B; exchange factor for ADP-ribosylation factor guanine nucleotide factor 6B; exchange factor for ARF6 B; PH and SEC7 domain-containing protein 4; pleckstrin and Sec7 domain containing 4; pleckstrin homology and SEC7 domain-containing protein 4; PSD4; SEC7 homolog; telomeric of interleukin-1 cluster protein; TIC
Gene Symbols PSD4
Host Species Rabbit
Immunogen A synthesized peptide derived from human PSD4(Accession Q8NDX1), corresponding to amino acid residues Q771-G821.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23550
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.