Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSD95 Monoclonal Antibody (6G6-1C9), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 56452100UG

This item is not returnable.
View return policy
Description
PSD95 Monoclonal antibody specifically detects PSD95 in Bovine, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
PSD95 | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P31016, Q62108 | |
Mouse | |
Protein A | |
RUO | |
100137840, 13385, 29495 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
6G6-1C9 | |
Unconjugated | |
DLG4 | |
discs large homolog 4; discs large MAGUK scaffold protein 4; discs, large homolog 4; discs, large homolog 4 (Drosophila); disks large homolog 4; Dlg4; Dlgh4; FLJ97752; FLJ98574; post synaptic density; postsynaptic density protein 95; post-synaptic density protein 95; PSD95; PSD-95; PSD-95 alpha 2b; PSD-95 beta; Sap90; SAP-90; SAP90A; Synapse-associated protein 90; synapse-associated protein SAP90; Tax interaction protein 15 | |
Recombinant rat PSD95 | |
100 μg | |
Primary | |
Bovine, Mouse, Rat | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction