Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PSEM 308 hydrochloride
GREENER_CHOICESDP

Catalog No. 642525 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-252-5 25 mg
64-255 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 64-252-5 Supplier Tocris Bioscience™ Supplier No. 6425/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

PSAML141F-GlyR and PSAML141F,Y115F-5-HT3 chimeric ion channel agonist

PSEM 308 hydrochloride is a PSAM (pharmacologically selective actuator module) agonist. Activates PSAML141F-GlyR chimeric ion channels. Inhibits activity of neurons expressing PSAML141F-GlyR in vivo and activates locus coeruleus noradrenergic neurons expressing PSAML141F,Y115F-5-HT3 ion channels.

Recommended concentration for use in mice is 5 mg/kg or lower.

Plasmid vectors for the transfection of cells with PSAML141F-GlyR and PSAML141F,Y115F-5-HT3 are available from Addgene.

Specifications

Chemical Name or Material 5-Methyl-5,8,9,10,11 A,12-hexahydro-4 H-8,11-ethanopyrido[3 ',2 ':3,4 ]diazepino[6,7,1-hi]indole hydrochloride
Quantity 25 mg
Target Pharmacologically Selective Effector Molecules/PSEM
Molecular Formula C17H21N3.HCl
Formula Weight 303.83
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.