Learn More
Description
Plasmid vectors for the transfection of cells with PSAML141F-GlyR and PSAML141F,Y115F-5-HT3 are available from Addgene.
Specifications
Specifications
| Chemical Name or Material | N-(3 S)-1-Azabicyclo[2.2.2 ]oct-3-yl-2,5-dimethoxybenzamide trifluoroacetate |
| CAS | 1336913-03-1 |
| Quantity | 25 mg |
| Target | Pharmacologically Selective Effector Molecules/PSEM |
| Molecular Formula | C16H22N2O3.CF3CO2H |
| Formula Weight | 404.38 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

