Learn More
Invitrogen™ PSEN2 Monoclonal Antibody (APS 26)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA1754
Description
MA1-754 detects presenilin 2 protein (PS2) from rat, mouse, human, and nonhuman primate samples. No cross-reactivity is seen with presenilin 1. MA1-754 has successfully been used in immuno-fluorescence, immunocytochemistry, Western blot and ELISA procedures. By Western blot, this antibody detects an ~20 kDa protein representing PS2 CT (C-terminal fragment) and the ~45 kDa full-length PS2 from transfected COS-7 cells. In 4% paraformaldehyde fixed RAW cells, MA1-754 showed specific Golgi and ER labeling. The MA1-754 immunogen is a synthetic peptide corresponding to residues C L(317) P Y D P E M E E D S Y D S F G E P(334) of human PS2.
Alzheimer's disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1 or PSEN2) or the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an enzyme that cleaves APP. Also, it is thought that the presenilins are involved in the cleavage of the Notch receptor such that, they either directly regulate gamma-secretase activity, or themselves act are protease enzymes. Two alternatively spliced transcript variants encoding different isoforms of PSEN2 have been identified.
Specifications
PSEN2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
O88777, P49810, Q61144 | |
PSEN2 | |
Synthetic peptide corresponding to residues C L(317) P Y D P E M E E D S Y D S F G E P(334) of human PS2. | |
200 μg | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
APS 26 | |
Unconjugated | |
PSEN2 | |
AD3L; AD3LP; AD4; Ad4h; AD5; Alg3; ALG-3; Alzheimer disease 4; CMD1V; E5-1; presenilin 2; presenilin-2; Presenilin-2 CTF subunit; Presenilin-2 NTF subunit; PS2; Ps-2; PSEN 2; PSEN2; Psnl2; STM2; STM-2 | |
Mouse | |
Protein G | |
RUO | |
19165, 5664, 81751 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.