Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSEN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112675
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112675 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112675 Supplier Thermo Scientific Supplier No. PA5112675
Add to Cart
Add to Cart

PSEN2 Polyclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen PSEN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PSEN2
Gene Accession No. O88777, P49810
Gene Alias AD3L; AD3LP; AD4; Ad4h; AD5; Alg3; ALG-3; Alzheimer disease 4; CMD1V; E5-1; presenilin 2; presenilin-2; Presenilin-2 CTF subunit; Presenilin-2 NTF subunit; PS2; Ps-2; PSEN 2; PSEN2; Psnl2; STM2; STM-2
Gene Symbols PSEN2
Host Species Rabbit
Immunogen Recombinant Human Presenilin-2 protein (7-77AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5664, 81751
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.